Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aerosol OT, 10% (w/v), Ricca Chemical
SDP

Catalog No. 28116 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
28-116 500 mL
281-32 1 L
281-1 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 28-116 Supplier Ricca Chemical Company Supplier No. 28116
Add to Cart
Add to Cart

Surfactant Solution

Specifications

Chemical Name or Material Aerosol OT, 10% (w/v)
Color Colorless to Yellow
CAS 7732-18-5, 64-17-5, 577-11-7, 67-63-0, 67-56-1
Physical Form Liquid
CAS Min % 73.74, 14.62, 10.1, 0.81, 0.73
Boiling Point ∼98°C
CAS Max % 73.74, 14.62, 10.1, 0.81, 0.73
Melting Point ∼0°C
Density 0.99
Concentration 10% (w/v)
Quantity 500 mL
Packaging Natural Poly Bottle
Solubility Information Miscible in water
Grade Laboratory
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.