Learn More
Labcon North America Labcon APT™ Aerosol Barrier Pipette Tips, Sterile, RNase/DNase Free, 4800 tips per case
Catalog No. NC3368298
Encompass_Preferred
The Labcon APT™ Aerosol Barrier Pipette Tips are sterile, 20 µL filter tips designed for contamination-free liquid handling in laboratories. With a unique aerosol barrier and compatibility with Beckman FX and NX AP96 Workstations, these tips prevent cross-contamination effectively. Made from high-quality materials and certified RNase/DNase free, they ensure reliable performance and precise results in scientific applications. Each case contains 4800 tips in autoclavable racks, supporting efficient workflow and enhanced accuracy.
Catalog No. NC3368298 Supplier Labcon North America Supplier No. 81146950009
The Labcon APT™ Aerosol Barrier Pipette Tips are sterile, 20 µL filter tips designed for contamination-free liquid handling in laboratories. With a unique aerosol barrier and compatibility with Beckman FX and NX AP96 Workstations, these tips prevent cross-contamination effectively. Made from high-quality materials and certified RNase/DNase free, they ensure reliable performance and precise results in scientific applications. Each case contains 4800 tips in autoclavable racks, supporting efficient workflow and enhanced accuracy.