Learn More
Justrite™ Aerosolv™ 7000 Super Aerosol Can Recycling System
Catalog No. 19121128 Shop All Justrite Safety Group Products
Change view
Click to view available options
Product Type:
Aerosol Can Recycling System
Catalog No. Product Type
19-121-128 Aerosol Can Recycling System
1 options
Catalog No. 19-121-128 Supplier Justrite™ Supplier No. 28230
  • $2,980.00
    $2,144.65 / Each of 1
    Save $835.35
    28% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart
Aerosolv™ 7000 Super aerosol can puncture device is a safe and efficient method for recycling spent aerosol cans
  • Offers all the features of original Aerosolv™ Standard System plus greater vapor reduction, easy tracking, and added security
  • Colormetric carbon filter indicates filter saturation
  • Lockout and shut-off mechanism seals system and drum when not in use
    • Eliminates escape of hazardous vapors to meet OSHA requirements
  • Easily comply with ISO 14001 program by using built-in counter
  • EPA compliant and ETV certified
  • Designed to eliminate entire RCRA waste stream from operations
  • Includes:
    • Combination coalescing/saturation indicating carbon filter
    • Padlock with key
    • Anti-static ground wire with C-clamp
    • Safety goggles
  • Specifications
    Certifications/Compliance ISO 14001, EPA compliant, ETV certified, OSHA requirements
    Weight (English) 9.75 lb.
    Weight (Metric) 4.42 kg
    Includes Combination coalescing/saturation indicating carbon filter, Padlock with key, Anti-static ground wire with C-clamp, Safety goggles
    Product Type Aerosol Can Recycling System