Learn More
Aerscher Diagnostics HemaPrompt™ Blood Test Kit
Catalog No. p-4418812
Click to view available options
For Use With (Application):
Fecal/Gastric Blood Test
Catalog No. For Use With (Application)
22-145-001 Fecal/Gastric Blood Test
1 options
Catalog No. 22-145-001 Supplier Aerscher Diagnostics Supplier No. HPFG
Add to Cart
Add to Cart
Guaiac-based combination occult blood test for fast and reliable results
  • Portability
  • Avoidance of cross-contamination
  • No developer bottle to lose, misplace, or misuse
  • No wait time to apply developer after specimen application
  • Results available in 1 minute
  • Standardized amount of developer solution
  • Only one item to stock
HemaPrompt FG: For fecal and Gastric occult blood test
HemaPrompt: For fecal Specimens only
Preliminary screening as an aid to diagnosis
Specifications
Clia Complexity Waived
Type HemaPrompt Blood Test Kit
For Use With (Application) Fecal/Gastric Blood Test
Quantity 50 Pack
Test Time 1 min.