Learn More
MSC Newstripe Aerosol Can Deflater, 30-55 Gal Drum, 1 ea
Catalog No. 502868711
Encompass
The Newstripe Inc. aerosol can deflater is designed for safe and efficient disposal of aerosol cans. It is compatible with both 30 and 55 gal drums, providing a comprehensive solution for industrial waste management.
  • Equipped with an anti-static cable for safety
  • Features a coalescing carbon filter
  • Includes an indicator check ball
  • Designed for use with 30 and 55 gal drums
  • Compact dimensions of 11 in high × 8 in wide × 30 in deep
Catalog No. 50-286-8711 Supplier MSC Supplier No. 33730128
The Newstripe Inc. aerosol can deflater is designed for safe and efficient disposal of aerosol cans. It is compatible with both 30 and 55 gal drums, providing a comprehensive solution for industrial waste management.
  • Equipped with an anti-static cable for safety
  • Features a coalescing carbon filter
  • Includes an indicator check ball
  • Designed for use with 30 and 55 gal drums
  • Compact dimensions of 11 in high × 8 in wide × 30 in deep