Learn More
Cayman Chemical AerugInascIn-d3 1mg
SDP
Supplier:  Cayman Chemical 371241 MG
Encompass_Preferred
An internal standard for the quantification of aeruginascin by GC- or LC-MS; intended for research and forensic applications
Catalog No. 50-291-5308