Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC AES-135 10mM 1mL | 2361659-61-0 | 1 ML
SDP

Catalog No. 5000229634
Encompass_Preferred

AES-135 is a hydroxamic acid-based pan-HDAC inhibitor supplied as a 10 mM solution in DMSO (1 mL) for research use only. It shows inhibitory activity against HDAC3, HDAC6, HDAC8, and HDAC11 and is provided with high reported purity and defined storage recommendations.

  • Hydroxamic acid-based pan-HDAC inhibitor.
  • Inhibits HDAC3, HDAC6, HDAC8, and HDAC11 (reported IC50s in sub-micromolar to low-micromolar range).
  • Provided as a 10 mM solution in DMSO, 1 mL.
  • Reported purity 98.21%.
  • Molecular formula C33H29F6N3O5S; molecular weight 693.66.
  • Storage recommendations: powder at -20°C (up to 3 years); solution at -80°C (up to 6 months).

Catalog No. 50-002-29634 Supplier Medchemexpress LLC Supplier No. HY11448310MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

AES-135 is a hydroxamic acid-based pan-HDAC inhibitor supplied as a 10 mM solution in DMSO (1 mL) for research use only. It shows inhibitory activity against HDAC3, HDAC6, HDAC8, and HDAC11 and is provided with high reported purity and defined storage recommendations.

  • Hydroxamic acid-based pan-HDAC inhibitor.
  • Inhibits HDAC3, HDAC6, HDAC8, and HDAC11 (reported IC50s in sub-micromolar to low-micromolar range).
  • Provided as a 10 mM solution in DMSO, 1 mL.
  • Reported purity 98.21%.
  • Molecular formula C33H29F6N3O5S; molecular weight 693.66.
  • Storage recommendations: powder at -20°C (up to 3 years); solution at -80°C (up to 6 months).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.