Learn More
Aviva Systems Biology Corp AES Antibody - middle region (ARP32323_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32323_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against AES. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human AES. Alternate names: AES-1, AES-2, ESP1, GRG, GRG5, TLE5ApplicationWB. Species reactivity: Cow, Guinea Pig, Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-54526