Learn More
Cayman Chemical aEthylamInopentIophenonhy 10mg
SDP
Catalog No. 502954550
Encompass_Preferred
A substituted cathinone that is an analog to ethcathinone, differing by having propyl rather than methyl at the α position; intended for forensic and research purposes.
Catalog No. 50-295-4550 Supplier Cayman Chemical Supplier No. 1428010 MG
A substituted cathinone that is an analog to ethcathinone, differing by having propyl rather than methyl at the α position; intended for forensic and research purposes.