Learn More
Cayman Chemical aEthylamInopentIophenonhy 50mg
SDP
Catalog No. 502954558
Encompass_Preferred
A substituted cathinone that is an analog to ethcathinone, differing by having propyl rather than methyl at the α position; intended for forensic and research purposes.
Catalog No. 50-295-4558 Supplier Cayman Chemical Supplier No. 1428050 MG
A substituted cathinone that is an analog to ethcathinone, differing by having propyl rather than methyl at the α position; intended for forensic and research purposes.