Learn More
Selleck Chemical LLC NVP-AEW541 5mg 475489-16-8 AEW541
SDP
Supplier:  Selleck Chemical LLC S10345MG
Encompass_Preferred
NVP-AEW541 is a potent inhibitor of IGF-1R/InsR with IC50 of 150 nM/140 nM in cell-free assays, greater potency and selectivity for IGF-1R in a cell-based assay. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0563537