Learn More
Selleck Chemical LLC AF-353 5mg
SDP
Supplier:  Selleck Chemical LLC S04055MG
Encompass_Preferred
AF-353 (Ro-4) is a potent, selective and orally bioavailable antagonist of P2X3 receptor and P2X2/3 receptor with pIC50 of 8.06, 8.05 and 7.41 for human P2X3, rat P2X3 and human P2X2/3, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3036