Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC AF-353 (Ro-4) | 865305-30-2 | 99.4% | C14H17IN4O2 | 1 ML
SDP

Catalog No. 5000354437
Encompass_Preferred

AF-353 (Ro-4) is a potent, selective, and orally bioavailable P2X3/P2X2/3 receptor antagonist. It inhibits α,β-meATP-evoked intracellular calcium flux and reduces the purinergic response in both normal and spinal cord-injured rats.

  • Potent and selective P2X3/P2X2/3 receptor antagonist
  • Orally bioavailable
  • Inhibits intracellular calcium flux
  • Reduces purinergic response in normal and SCI rats
  • Does not compromise oxygen levels or cardiac function

Catalog No. 50-003-54437 Supplier Medchemexpress LLC Supplier No. HY1448310MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

AF-353 (Ro-4) is a potent, selective, and orally bioavailable P2X3/P2X2/3 receptor antagonist. It inhibits α,β-meATP-evoked intracellular calcium flux and reduces the purinergic response in both normal and spinal cord-injured rats.

  • Potent and selective P2X3/P2X2/3 receptor antagonist
  • Orally bioavailable
  • Inhibits intracellular calcium flux
  • Reduces purinergic response in normal and SCI rats
  • Does not compromise oxygen levels or cardiac function

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.