Learn More
Jackson Immuno Research Labs Alexa Fluor 594 AffiniPure Goat Anti-Rabbit IgG (H+L), 1mg
SDP
Supplier:  Jackson Immuno Research Labs 111585144
Encompass_Preferred
aF 594aFFINIPURE GOaT aNTI-RaB. Whole IgG antibodies are isolated as intact molecules from antisera by immunoaffinity chromatography. They have an Fc portion and two antigen binding Fab portions joined together by disulfide bonds and therefore they are divalent. The average molecular weight is reported to be about 160 kDa. The whole IgG form of antibodies is suitable for the majority of immunodetection procedures and is the most cost effective. Based on immunoelectrophoresis and/or ELISa, the antibody reacts with whole molecule rabbit IgG. It also reacts with the light chains of other rabbit immunoglobulins. No antibody was detected against non-immunoglobulin serum proteins. The antibody has been tested by ELISa and/or solid-phase adsorbed to ensure minimal cross-reaction with human, mouse and rat serum proteins, but it may cross-react with immunoglobulins from other species.
Catalog No. NC0236158