Learn More
Biolegend Alexa Fluor 647 anti-mouse H-2Dd
Supplier:  Biolegend 110612
Encompass_Preferred
Alexa Fluor 647 anti-mouse H-2Dd [34-2-12]; Isotype: Mouse (C3H/HeJ) IgG2a, κ; Reactivity: Mouse d haplotype, Cross-reacts with the C3H.LG/Ckc; Apps: FC; Size: 100 ug
Catalog No. NC1791510