Learn More
Biolegend Alexa Fluor 647 anti-human CD340 (erbB2/HER-2)
Supplier:  Biolegend 324412
Encompass_Preferred
Alexa Fluor 647 anti-human CD340 (erbB2/HER-2) [24D2]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC, ICC; Size: 100 tests
Catalog No. NC1573463