Learn More
Biolegend Alexa Fluor 700 anti-mouse CD69
Supplier:  Biolegend 104539
Encompass_Preferred
Alexa Fluor 700 anti-mouse CD69 [H1.2F3]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. NC1022766