Learn More
Sigma Aldrich Fine Chemicals Biosciences Af HDL and LDL/VLDL Assay
Catalog No. 502904262
Encompass
Cholesterol concentrations in High-Density Lipoprotein (HDL) and Low-Density (LDL)/Very-Low-Density (VLDL) Lipoproteins are strong predictors for coronary heart disease. functional HDL offers protection by removing cholesterol from cells and atheroma. Higher concentrations of LDL and lower concentrations of functional HDL are strongly associated with cardiovascular disease due to higher risk of atherosclerosis. The balances between high- and low-density lipoproteins are solely genetically determined but can be changed by medications food choices and other factors. Simple direct and automation-ready procedures for measuring HDL and LDL/VLDL concentrations are very desirable.
Catalog No. 50-290-4262 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. MAK3311KT
Cholesterol concentrations in High-Density Lipoprotein (HDL) and Low-Density (LDL)/Very-Low-Density (VLDL) Lipoproteins are strong predictors for coronary heart disease. functional HDL offers protection by removing cholesterol from cells and atheroma. Higher concentrations of LDL and lower concentrations of functional HDL are strongly associated with cardiovascular disease due to higher risk of atherosclerosis. The balances between high- and low-density lipoproteins are solely genetically determined but can be changed by medications food choices and other factors. Simple direct and automation-ready procedures for measuring HDL and LDL/VLDL concentrations are very desirable.