Learn More
MSC Intertape Aluminum Foil Tape, 4.7 mil, 1 in, 1 ea
Catalog No. 501304706
Encompass
The ALF300 series aluminum foil tape features a 3.0 mil dead soft aluminum foil backing with an aggressive pressure-sensitive acrylic adhesive. It offers instant, water-resistant adhesion to clean, dry surfaces, and dispenses easily from the roll. This tape conforms to irregular and curved surfaces, maintains flexibility, and resists cracking and lifting, making it suitable for various applications.
  • Backing of 3.0 mil dead soft aluminum foil
  • Aggressive pressure-sensitive acrylic adhesive
  • Instant, water-resistant adhesion
  • Adheres well to itself
  • Dispenses easily from the roll
  • Conforms to irregular/curved surfaces
  • Remains flexible and resists cracking and lifting
  • UV resistant
Catalog No. 50-130-4706 Supplier MSC Supplier No. 62472592
The ALF300 series aluminum foil tape features a 3.0 mil dead soft aluminum foil backing with an aggressive pressure-sensitive acrylic adhesive. It offers instant, water-resistant adhesion to clean, dry surfaces, and dispenses easily from the roll. This tape conforms to irregular and curved surfaces, maintains flexibility, and resists cracking and lifting, making it suitable for various applications.
  • Backing of 3.0 mil dead soft aluminum foil
  • Aggressive pressure-sensitive acrylic adhesive
  • Instant, water-resistant adhesion
  • Adheres well to itself
  • Dispenses easily from the roll
  • Conforms to irregular/curved surfaces
  • Remains flexible and resists cracking and lifting
  • UV resistant