Learn More
Thermo Scientific AS19-4µm IC Columns
Catalog No. NC0661134 Shop All Thermo Scientific Products
Encompass
Achieve high resolution separations of trace oxyhalides and inorganic anions in drinking water using the Thermo Scientific™ Dionex™ IonPac™ AS19-4µm hydroxide-selective anion-exchange column. Standard Bore Guard 4mm x 250mm. Flow Rate: 1.0 to 1.5mL/min. pH 0-14; 0-100% HPLC Solvents.
Catalog No. NC0661134 Supplier Thermo Scientific Supplier No. 083217
Achieve high resolution separations of trace oxyhalides and inorganic anions in drinking water using the Thermo Scientific™ Dionex™ IonPac™ AS19-4µm hydroxide-selective anion-exchange column. Standard Bore Guard 4mm x 250mm. Flow Rate: 1.0 to 1.5mL/min. pH 0-14; 0-100% HPLC Solvents.