Learn More
Medchemexpress LLC HY-12802 5mg Medchemexpress, AF38469 CAS:1531634-31-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY128025MG
Encompass_Preferred
Medchemexpress, HY-12802 5mg AF38469 CAS:1531634-31-7 AF38469 is a selective, orally bioavailable Sortilin inhibitor with an IC50 value of 330 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8916