Learn More
Medchemexpress LLC HY-124673 5mg Medchemexpress, AF40431 CAS:181125-92-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1246735MG
Encompass_Preferred
Medchemexpress, HY-124673 5mg AF40431 CAS:181125-92-8 AF40431, the first reported small-molecule ligand of sortilin, has an IC50 of 4.4 µM and a Kd of 0.7 µM . AF40431 is bound in the neurotensin-binding site of sortilin. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9676