Learn More
Lumiprobe AF488 NHS ester, 5 mg
Catalog No. NC1863173
Encompass_Preferred
AF488 is a bright and photostable dye, equivalent of Alexa Fluor 488. Due to its high hydrophilicity, this is a dye of choice for the labeling of sensitive proteins and antibodies. The dye is useful for many demanding applications, including microscopy.
Catalog No. NC1863173 Supplier Lumiprobe Supplier No. 218205MG
AF488 is a bright and photostable dye, equivalent of Alexa Fluor 488. Due to its high hydrophilicity, this is a dye of choice for the labeling of sensitive proteins and antibodies. The dye is useful for many demanding applications, including microscopy.