Learn More
Jackson Immuno Research Labs Alexa Fluor 647 AffiniPure Alpaca Anti-Rabbit IgG (H+L) | 1.0mg
SDP
Catalog No. NC1464759
Encompass_Preferred
Target: Rabbit | Host: Alpaca | Antibody Format: Whole IgG | Specificity: IgG (H+L) | Minimal Cross Reactivity: Bovine, Human, Mouse Serum Proteins | Conjugate: Alexa Fluor 647
Catalog No. NC1464759 Supplier Jackson Immuno Research Labs Supplier No. 611605215
Add to Cart
Add to Cart
Target: Rabbit | Host: Alpaca | Antibody Format: Whole IgG | Specificity: IgG (H+L) | Minimal Cross Reactivity: Bovine, Human, Mouse Serum Proteins | Conjugate: Alexa Fluor 647