Learn More
Lumiprobe AF 647 NHS Ester, 5 mg, far-red fluorescent dye, NHS ester, pH-insensitive (4-10), high water solubility, applications: cellular visualization, imaging, flow cytometry
Catalog No. NC2143481
Encompass_Preferred
AF 647 NHS Ester, 5 mg, far-red fluorescent dye, NHS ester, pH-insensitive (4-10), high water solubility, applications: cellular visualization, imaging, flow cytometry
This item is not returnable. View return policy
Catalog No. NC2143481 Supplier Lumiprobe Supplier No. 26820
AF 647 NHS Ester, 5 mg, far-red fluorescent dye, NHS ester, pH-insensitive (4-10), high water solubility, applications: cellular visualization, imaging, flow cytometry