Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC AF647-NHS ester (triTEA) | 407627-61-6 | 99.7% | 10 MG
SDP

Catalog No. 5000297841
Encompass_Preferred

AF647-NHS ester (triTEA) is a far-red fluorescent NHS ester reagent used to label primary amines for fluorescence detection. Supplied as a purple solid, it is optimized for conventional and instantaneous detection and is soluble in DMSO when warmed or sonicated.

  • Far-red fluorescent NHS ester for amine conjugation.
  • Excitation: 635 nm (conventional) / 620 nm (instantaneous).
  • Physical form: purple to dark purple solid.
  • Purity: 99.74% as reported by manufacturer.
  • Solubility: DMSO 20 mg/mL; use freshly opened DMSO and apply ultrasonic/heat to 60°C as needed.
  • Storage: protect from light and store under nitrogen; in solvent: -80°C (6 months), -20°C (1 month).
  • For research use only; not for patient use.

Catalog No. 50-002-97841 Supplier Medchemexpress LLC Supplier No. HYD1119C10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

AF647-NHS ester (triTEA) is a far-red fluorescent NHS ester reagent used to label primary amines for fluorescence detection. Supplied as a purple solid, it is optimized for conventional and instantaneous detection and is soluble in DMSO when warmed or sonicated.

  • Far-red fluorescent NHS ester for amine conjugation.
  • Excitation: 635 nm (conventional) / 620 nm (instantaneous).
  • Physical form: purple to dark purple solid.
  • Purity: 99.74% as reported by manufacturer.
  • Solubility: DMSO 20 mg/mL; use freshly opened DMSO and apply ultrasonic/heat to 60°C as needed.
  • Storage: protect from light and store under nitrogen; in solvent: -80°C (6 months), -20°C (1 month).
  • For research use only; not for patient use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.