Learn More
Medchemexpress LLC Af647-nhs ester tritea | 407627-61-6 | 99.7% | 1259.66 | 5 MG
SDP
Catalog No. 5000428173
Encompass_Preferred
AF647-NHS ester is an analogue of Alexa Fluor 647 (AF647) with an excitation wavelength (λex) of 635 nm (conventional fluorescence detection) / 620 nm (instantaneous detection). The NHS ester can covalently bind to molecules with amino groups, such as proteins and antibodies.
  • Analogue of Alexa Fluor 647 (AF647)
  • Excitation wavelength (λex) of 635 nm (conventional fluorescence detection) / 620 nm (instantaneous detection)
  • NHS ester can covalently bind to molecules with amino groups (e.g., proteins, antibodies)
Catalog No. 50-004-28173 Supplier Medchemexpress LLC Supplier No. HYD1119C5MG
AF647-NHS ester is an analogue of Alexa Fluor 647 (AF647) with an excitation wavelength (λex) of 635 nm (conventional fluorescence detection) / 620 nm (instantaneous detection). The NHS ester can covalently bind to molecules with amino groups, such as proteins and antibodies.
  • Analogue of Alexa Fluor 647 (AF647)
  • Excitation wavelength (λex) of 635 nm (conventional fluorescence detection) / 620 nm (instantaneous detection)
  • NHS ester can covalently bind to molecules with amino groups (e.g., proteins, antibodies)