Learn More
Medchemexpress LLC AF647-NHS ester (triTEA) | 407627-61-6 | 99.74% | 1259.66 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYD1119C
Encompass_Preferred
AF647-NHS ester is an analogue of Alexa Fluor 647, which has an excitation wavelength (λex) of 635 nm (conventional fluorescence detection) / 620 nm (instantaneous detection). The NHS ester functional group allows it to covalently bind to molecules with amino groups such as proteins and antibodies.
  • Analogue of Alexa Fluor 647
  • Excitation wavelength (λex) of 635 nm (conventional fluorescence detection) / 620 nm (instantaneous detection)
  • Features an NHS ester functional group
  • Covalently binds to molecules with amino groups
  • For research use only
Catalog No. 50-004-29520