Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC AF680-conjugated Donkey Anti-Goat IgG H&L | 100 UL
SDP

Catalog No. 5000443600
Encompass_Preferred

AF680-conjugated Donkey Anti-Goat IgG H&L is a red AF680-conjugated and donkey-originated polyclonal antibody, targeting goat IgG antibody. It can bind to the light and heavy chains of goat IgG antibodies.

  • Host is donkey
  • Clonality is polyclonal
  • Reacts with goat species
  • Immunogen is goat IgG
  • Affinity purified
  • Conjugated with AF 680
  • Isotype is IgG
  • Appears as liquid
  • Supplied in 0.01M Phosphate Buffered Saline, pH 7.2, containing 1% BSA, 50% glycerol, 0.02% Sodium Azide
  • Stored at -20°C for 1 year. Protect from light. Avoid repeated freeze/thaw cycles

Catalog No. 50-004-43600 Supplier Medchemexpress LLC Supplier No. HYP83744100UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

AF680-conjugated Donkey Anti-Goat IgG H&L is a red AF680-conjugated and donkey-originated polyclonal antibody, targeting goat IgG antibody. It can bind to the light and heavy chains of goat IgG antibodies.

  • Host is donkey
  • Clonality is polyclonal
  • Reacts with goat species
  • Immunogen is goat IgG
  • Affinity purified
  • Conjugated with AF 680
  • Isotype is IgG
  • Appears as liquid
  • Supplied in 0.01M Phosphate Buffered Saline, pH 7.2, containing 1% BSA, 50% glycerol, 0.02% Sodium Azide
  • Stored at -20°C for 1 year. Protect from light. Avoid repeated freeze/thaw cycles

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.