Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC AF680-conjugated Donkey Anti-Goat IgG H&L | 50 UL
SDP

Catalog No. 5000444183
Encompass_Preferred

AF680-conjugated Donkey Anti-Goat IgG H&L is a red AF680-conjugated and Donkey origined Polyclonal antibody, targeting Goat IgG antibody. It binds to the light and heavy chains of Goat IgG antibodies. This product is for research use only.

  • Host: Donkey
  • Clonality: Polyclonal
  • Species reactivity: Goat
  • Immunogen: Goat IgG
  • Purity: Affinity purified
  • Conjugation: AF 680
  • Isotype: IgG
  • Appearance: Liquid
  • Formulation: Supplied in 0.01M Phosphate Buffered Saline, pH 7.2, containing 1% BSA, 50% glycerol, 0.02% Sodium Azide.
  • Storage: Stored at -20°C for 1 year. Protect from light. Avoid repeated freeze/thaw cycles.

Catalog No. 50-004-44183 Supplier Medchemexpress LLC Supplier No. HYP8374450UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

AF680-conjugated Donkey Anti-Goat IgG H&L is a red AF680-conjugated and Donkey origined Polyclonal antibody, targeting Goat IgG antibody. It binds to the light and heavy chains of Goat IgG antibodies. This product is for research use only.

  • Host: Donkey
  • Clonality: Polyclonal
  • Species reactivity: Goat
  • Immunogen: Goat IgG
  • Purity: Affinity purified
  • Conjugation: AF 680
  • Isotype: IgG
  • Appearance: Liquid
  • Formulation: Supplied in 0.01M Phosphate Buffered Saline, pH 7.2, containing 1% BSA, 50% glycerol, 0.02% Sodium Azide.
  • Storage: Stored at -20°C for 1 year. Protect from light. Avoid repeated freeze/thaw cycles.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.