Learn More
Biolegend Alexa Fluor 700 anti-human CD45
Supplier:  Biolegend 368513
Encompass_Preferred
Alexa Fluor 700 anti-human CD45 [2D1]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. NC1359040