Learn More
Biolegend Alexa Fluor 700 anti-human CD123
Supplier:  Biolegend 306040
Encompass_Preferred
Alexa Fluor 700 anti-human CD123 [6H6]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Rhesus; Apps: FC; Size: 100 tests
Catalog No. NC1630342