Learn More
Zymo Research Corporation a-Factor Mating Pheromone, 500 ul
SDP
Supplier:  Zymo Research Corporation Y1004500
Encompass
a-factor is one of the two mating pheromones in baking yeast. It is the opposite sex of mating pheromone alpha-Factor (alpha-factor). a-Factor is functionally tested for its activity and is stable for multiple freeze-thaw cycles.
Catalog No. NC0924426