Learn More
Medchemexpress LLC Afalanine (N-Acetyl-DL-phenylalanine) | 2901-75-9 | 99.9% | 200 MG
SDP
Catalog No. 5000413769
Encompass_Preferred
Afalanine (N-Acetyl-DL-phenylalanine) is an endogenous metabolite of an endophytic fungus with antidepressant activity. It can be used in combination with antibiotics to prevent renal damage.
  • Endogenous metabolite of an endophytic fungus
  • Antidepressant activity
  • Can be used in combination with antibiotics to prevent renal damage
  • For research use only
Catalog No. 50-004-13769 Supplier Medchemexpress LLC Supplier No. HYB1086200MG
Afalanine (N-Acetyl-DL-phenylalanine) is an endogenous metabolite of an endophytic fungus with antidepressant activity. It can be used in combination with antibiotics to prevent renal damage.
  • Endogenous metabolite of an endophytic fungus
  • Antidepressant activity
  • Can be used in combination with antibiotics to prevent renal damage
  • For research use only