Learn More
TARGETMOL CHEMICALS INC Afamelanotide acetate 5MG
SDP
Catalog No. 502878027
Encompass_Preferred
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Afamelanotide acetate is a synthetic alpha-melanocyte stimulating hormone analogue and first-in-class melanocortin-1 receptor agonist, used to for Erythropoietic Protoporphyria. Purity 98%
Catalog No. 50-287-8027 Supplier TARGETMOL CHEMICALS INC Supplier No. T75895MG
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Afamelanotide acetate is a synthetic alpha-melanocyte stimulating hormone analogue and first-in-class melanocortin-1 receptor agonist, used to for Erythropoietic Protoporphyria. Purity 98%