Learn More
Aviva Systems Biology Corp AFAP1L2 Antibody - N-terminal region (ARP60221_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60221_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against AFAP1L2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human AFAP1L2. Alternate names: FLJ14564, KIAA1914, XB130ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-978