Learn More
Medchemexpress LLC HY-10261 200mg , Afatinib BIBW 2992 CAS:850140-72-6 Purity:98%
SDP
Supplier:  Medchemexpress LLC HY10261200MG
Encompass_Preferred
Medchemexpress, HY-10261 200mg Afatinib BIBW 2992 CAS:850140-72-6 Formula:C24H25ClFN5O3 IC50: 0.5 nM (EGFR wt ), 0.4 nM (EGFR L858R ), 10 nM (EGFR L858R/T790M ), 14 nM (HER2) Purity:98% Afatinib is an irreversible, dual EGFR / HER2 inhibitor, shows potent activity against wild-type and mutant forms of EGFR and HER2, with IC 50 of 0.5 nM, 0.4 nM, 10 nM and 14 nM for EGFR wt , EGFR L858R , EGFR L858R/T790M and HER2, respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2522