Learn More
Selleck Chemical LLC Afatinib S1011-200MG
SDP
Catalog No. 5000038906
Encompass_Preferred
Afatinib inhibits EGFR/ErbB irreversibly in vitro with IC50 of 0 5 0 4 10 14 1 nM for EGFRwt EGFR L858R EGFR L858R/T790M ErbB2 (HER2) and ErbB4 (HER4) respectively Afatinib induces autophagy
Catalog No. 50-000-38906 Supplier Selleck Chemical LLC Supplier No. S1011200MG
Afatinib inhibits EGFR/ErbB irreversibly in vitro with IC50 of 0 5 0 4 10 14 1 nM for EGFRwt EGFR L858R EGFR L858R/T790M ErbB2 (HER2) and ErbB4 (HER4) respectively Afatinib induces autophagy