Learn More
Apexbio Technology LLC Afatinib (BIBW2992) 439081-18-2 10mM (in 1mL DMSO)
SDP
Catalog No. 5000061587
Encompass_Preferred
Afatinib (BIBW2992 CAS 439081-18-2) is an irreversible inhibitor targeting receptor tyrosine kinases in the ErbB protein family specifically EGFR (ErbB1) and HER2 (ErbB2) By covalently binding to their kinase domains it suppresses downstream ErbB signaling showing demonstrated inhibition with IC50 values of 0 5 nM for EGFR and 14 nM for HER2 Afatinib blocks EGF-induced phosphorylation and cellular proliferation in multiple EGFR- or HER2-expressing lines including A431 NIH-3T3-HER2 NCI-N87 and BT-474 Research applications include tumor xenograft studies and transgenic lung cancer models evaluating EGFR-dependent oncogenic processes and therapeutic responses
Catalog No. 50-000-61587 Supplier Apexbio Technology LLC Supplier No. A82475.1
Afatinib (BIBW2992 CAS 439081-18-2) is an irreversible inhibitor targeting receptor tyrosine kinases in the ErbB protein family specifically EGFR (ErbB1) and HER2 (ErbB2) By covalently binding to their kinase domains it suppresses downstream ErbB signaling showing demonstrated inhibition with IC50 values of 0 5 nM for EGFR and 14 nM for HER2 Afatinib blocks EGF-induced phosphorylation and cellular proliferation in multiple EGFR- or HER2-expressing lines including A431 NIH-3T3-HER2 NCI-N87 and BT-474 Research applications include tumor xenograft studies and transgenic lung cancer models evaluating EGFR-dependent oncogenic processes and therapeutic responses