Learn More
Selleck Chemical LLC Afatinib (BIBW2992) 10mM/1mL 850140-72-6
SDP
Supplier:  Selleck Chemical LLC S101110MM/1ML
Encompass_Preferred
Afatinib (BIBW2992) inhibits EGFR/ErbB irreversibly in vitro with IC50 of 0.5, 0.4, 10, 14, 1 nM for EGFRwt, EGFR L858R , EGFR L858R/T790M ErbB2 (HER2) and ErbB4 (HER4), respectively. Afatinib induces autophagy. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-0856