Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Afatinib-d6 | 1313874-96-2 | 99.0% | 491.98 | C24H19D6ClFN5O3 | 10 MG
SDP

Catalog No. 5000245450
Encompass_Preferred

Afatinib-d6 is a deuterium-labeled analogue of the irreversible EGFR-family inhibitor afatinib, provided for research use as an internal standard or tracer in quantitative analytical methods (LC-MS, GC-MS, NMR). It is intended for analytical and pharmacokinetic studies where accurate quantification is required.

  • Deuterium-labeled internal standard for afatinib quantification.
  • High deuterium incorporation; purity ≥99.0%.
  • Molecular formula C24H19D6ClFN5O3; molecular weight 491.98.
  • Supplied as a 10 mg package suitable for analytical assays.
  • For research use only; not for human or clinical use.

Catalog No. 50-002-45450 Supplier Medchemexpress LLC Supplier No. HY10261S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Afatinib-d6 is a deuterium-labeled analogue of the irreversible EGFR-family inhibitor afatinib, provided for research use as an internal standard or tracer in quantitative analytical methods (LC-MS, GC-MS, NMR). It is intended for analytical and pharmacokinetic studies where accurate quantification is required.

  • Deuterium-labeled internal standard for afatinib quantification.
  • High deuterium incorporation; purity ≥99.0%.
  • Molecular formula C24H19D6ClFN5O3; molecular weight 491.98.
  • Supplied as a 10 mg package suitable for analytical assays.
  • For research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.