Learn More
Medchemexpress LLC Afatinib-d6 | 1313874-96-2 | 99.0% | 491.98 g/mol | C24H19D6ClFN5O3 | 1 MG
SDP
Catalog No. 5000245451
Encompass_Preferred
Afatinib-d6 is the deuterium-labeled isotopologue of afatinib, an irreversible ErbB (EGFR/HER2) family inhibitor. Provided as a high-purity analytical standard, it is intended for research applications including mass spectrometry, pharmacokinetic studies, and tracer experiments. CAS 1313874-96-2; molecular weight 491.98 g/mol; chemical formula C24H19D6ClFN5O3.
  • Deuterium-labeled internal standard for mass spectrometry
  • High purity (99.0%) suitable for analytical use
  • Useful in pharmacokinetic and metabolic studies
  • Irreversible ErbB (EGFR/HER2) inhibitor scaffold
  • Available in small research quantities for method development
Catalog No. 50-002-45451 Supplier Medchemexpress LLC Supplier No. HY10261S1MG
Add to Cart
Add to Cart
Afatinib-d6 is the deuterium-labeled isotopologue of afatinib, an irreversible ErbB (EGFR/HER2) family inhibitor. Provided as a high-purity analytical standard, it is intended for research applications including mass spectrometry, pharmacokinetic studies, and tracer experiments. CAS 1313874-96-2; molecular weight 491.98 g/mol; chemical formula C24H19D6ClFN5O3.
  • Deuterium-labeled internal standard for mass spectrometry
  • High purity (99.0%) suitable for analytical use
  • Useful in pharmacokinetic and metabolic studies
  • Irreversible ErbB (EGFR/HER2) inhibitor scaffold
  • Available in small research quantities for method development