Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Afatinib-d6 | 1313874-96-2 | 99.0% | 491.98 g/mol | C24H19D6ClFN5O3 | 5 MG
SDP

Catalog No. 5000246969
Encompass_Preferred

Afatinib-d6 is a deuterium-labeled isotopologue of afatinib supplied as a high-purity analytical reference for use in LC-MS/MS, pharmacokinetic studies, and other research requiring an internal standard. It is intended for analytical and research use where isotopic labeling improves quantitation and identification.

  • Deuterium-labeled afatinib for use as an internal standard.
  • Irreversible EGFR family inhibitor analog compatible with LC-MS/MS workflows.
  • High purity (99.0%) suitable for analytical applications.
  • Molecular weight 491.98 g/mol; formula C24H19D6ClFN5O3; CAS 1313874-96-2.
  • Supplied in small quantities for analytical use (example: 5 mg vial).

Catalog No. 50-002-46969 Supplier Medchemexpress LLC Supplier No. HY10261S5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Afatinib-d6 is a deuterium-labeled isotopologue of afatinib supplied as a high-purity analytical reference for use in LC-MS/MS, pharmacokinetic studies, and other research requiring an internal standard. It is intended for analytical and research use where isotopic labeling improves quantitation and identification.

  • Deuterium-labeled afatinib for use as an internal standard.
  • Irreversible EGFR family inhibitor analog compatible with LC-MS/MS workflows.
  • High purity (99.0%) suitable for analytical applications.
  • Molecular weight 491.98 g/mol; formula C24H19D6ClFN5O3; CAS 1313874-96-2.
  • Supplied in small quantities for analytical use (example: 5 mg vial).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.