Learn More
Medchemexpress LLC HY-10261A 10mg Medchemexpress, Afatinib (dimaleate) CAS:850140-73-7 Purity:>98%
SDP
Catalog No. 501967993
Encompass_Preferred
Medchemexpress, HY-10261A 10mg Afatinib (dimaleate) CAS:850140-73-7 Afatinib dimaleate is an irreversible EGFR family inhibitor with IC50s of 0.5 nM, 0.4 nM, 10 nM and 14 nM for EGFRwt, EGFRL858R, EGFRL858R/T790M and HER2, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7993 Supplier Medchemexpress LLC Supplier No. HY10261A10MG
Medchemexpress, HY-10261A 10mg Afatinib (dimaleate) CAS:850140-73-7 Afatinib dimaleate is an irreversible EGFR family inhibitor with IC50s of 0.5 nM, 0.4 nM, 10 nM and 14 nM for EGFRwt, EGFRL858R, EGFRL858R/T790M and HER2, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.