Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Afatinib dimaleate | 850140-73-7 | MFCD25974239 | >98.0% | 718.08 g/mol | C32H33ClFN5O11 | 100 MG
SDP

Catalog No. 5000245452
Encompass_Preferred

Afatinib dimaleate is the dimaleate salt form of afatinib, an orally active, irreversible inhibitor of ErbB family tyrosine kinases (EGFR and HER2). It is supplied for research and analytical applications with defined purity and storage recommendations.

  • Potent, irreversible inhibitor of EGFR and HER2.
  • Provided as the dimaleate salt for improved handling and stability.
  • Reported high purity (>98% by HPLC) suitable for research use.
  • Molecular weight 718.08 g/mol.
  • Recommended storage: sealed and away from moisture; in solvent -80°C (6 months), -20°C (1 month).

Catalog No. 50-002-45452 Supplier Medchemexpress LLC Supplier No. HY10261A100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Afatinib dimaleate is the dimaleate salt form of afatinib, an orally active, irreversible inhibitor of ErbB family tyrosine kinases (EGFR and HER2). It is supplied for research and analytical applications with defined purity and storage recommendations.

  • Potent, irreversible inhibitor of EGFR and HER2.
  • Provided as the dimaleate salt for improved handling and stability.
  • Reported high purity (>98% by HPLC) suitable for research use.
  • Molecular weight 718.08 g/mol.
  • Recommended storage: sealed and away from moisture; in solvent -80°C (6 months), -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.