Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Afatinib (E/Z isomer mixture) | 439081-18-2 | MFCD12407405 | 100.0% | 485.94 g/mol | C24H25ClFN5O3 | 100 MG
SDP

Catalog No. 5000248403
Encompass_Preferred

(E/Z)-Afatinib is a mixture of the E and Z isomers of afatinib, an irreversible tyrosine kinase inhibitor that targets the ErbB family (EGFR, HER2, HER4, and EGFRvIII). Supplied as a solid for research use, it is commonly used in biochemical and cell-based studies to inhibit EGFR signaling.

  • Mixture of E and Z isomers.
  • Irreversible inhibitor of EGFR, HER2, HER4, and EGFRvIII.
  • Provided as a solid suitable for formulation or assay preparation.
  • Typical pack size: 100 MG solid.
  • Chemical formula: C24H25ClFN5O3; molecular weight ≈ 485.94 g/mol.

Catalog No. 50-002-48403 Supplier Medchemexpress LLC Supplier No. HY10261B100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(E/Z)-Afatinib is a mixture of the E and Z isomers of afatinib, an irreversible tyrosine kinase inhibitor that targets the ErbB family (EGFR, HER2, HER4, and EGFRvIII). Supplied as a solid for research use, it is commonly used in biochemical and cell-based studies to inhibit EGFR signaling.

  • Mixture of E and Z isomers.
  • Irreversible inhibitor of EGFR, HER2, HER4, and EGFRvIII.
  • Provided as a solid suitable for formulation or assay preparation.
  • Typical pack size: 100 MG solid.
  • Chemical formula: C24H25ClFN5O3; molecular weight ≈ 485.94 g/mol.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.