Learn More
Diagnostic Biosystems AFB AFB
Catalog No. 502407599
Encompass_Preferred
This kit stains acid-fast bacteria and tubercle bacilli of Mycobacterium genus in paraffin tissue sections. Tubercle bacilli and other acid-fast bacteria stain bright red and the background stains light green.
Catalog No. 50-240-7599 Supplier Diagnostic Biosystems Supplier No. KT001
This kit stains acid-fast bacteria and tubercle bacilli of Mycobacterium genus in paraffin tissue sections. Tubercle bacilli and other acid-fast bacteria stain bright red and the background stains light green.