Learn More
Newcomer Supply AFB (Acid Fast Bacteria), Basic Fuchsin, Ellis & Zabrowarny Stain Kit, 250 ml kit
SDP
Catalog No. NC2102252
Encompass
Newcomer Supply AFB, Basic Fuchsin, Ellis & Zabrowarny Stain Kit procedure, with included microwave modification, provides a safe method for staining acid-fast bacilli without the use of phenol.  Results are comparable to the AFB, Ziehl-Neelsen method.
This item is not returnable. View return policy
Catalog No. NC2102252 Supplier Newcomer Supply Supplier No. 91012A
Newcomer Supply AFB, Basic Fuchsin, Ellis & Zabrowarny Stain Kit procedure, with included microwave modification, provides a safe method for staining acid-fast bacilli without the use of phenol.  Results are comparable to the AFB, Ziehl-Neelsen method.