Learn More
Ethos Biosciences AFB Brilliant Green Kinyoun
Catalog No. 5000819180
Encompass_Preferred
AFB Brilliant Green Counterstain is designed for use in acid-fast staining procedures including the Kinyoun method to provide strong contrast between acid-fast organisms and background material When used after carbol fuchsin staining and decolorization the counterstain produces a clean green background allowing acid-fast bacteria to appear bright red for clear visualization Suitable for routine use in clinical veterinary and research laboratories this reagent is compatible with standard acid-fast staining protocols and manual procedures The formulation supports consistent color development and reliable results in microbiology applications
Catalog No. 50-008-19180 Supplier Ethos Biosciences Supplier No. 630308
AFB Brilliant Green Counterstain is designed for use in acid-fast staining procedures including the Kinyoun method to provide strong contrast between acid-fast organisms and background material When used after carbol fuchsin staining and decolorization the counterstain produces a clean green background allowing acid-fast bacteria to appear bright red for clear visualization Suitable for routine use in clinical veterinary and research laboratories this reagent is compatible with standard acid-fast staining protocols and manual procedures The formulation supports consistent color development and reliable results in microbiology applications