Learn More
Newcomer Supply Acid Fast Bacteria (AFB), Animal Control Slides, 10/set
SDP
Catalog No. NC9308274
Encompass
Newcomer Supply Acid Fast Bacteria (AFB), Animal Control Slides are for the positive histochemical staining of acid-fast mycobacteria in tissue sections. Acid Fast Bacteria (AFB), Animal Control Slides contain sections of positive staining animal organ and negative staining human lung.
Catalog No. NC9308274 Supplier Newcomer Supply Supplier No. 4011A
Newcomer Supply Acid Fast Bacteria (AFB), Animal Control Slides are for the positive histochemical staining of acid-fast mycobacteria in tissue sections. Acid Fast Bacteria (AFB), Animal Control Slides contain sections of positive staining animal organ and negative staining human lung.